Silver nitrite, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Argentous nitrite
M-Clarity™ quality level = MQ100
545015-25G
545015-5G
SIAL545015-25GEA
0
CZK
SIAL545015-25G
SIAL545015-5G
Silver nitrite, Sigma-Aldrich®
Silver nitrite
Vzorec:
AgNO₂ MW: 153,87 g/mol Bod tání: 140 °C Hustota: 4,45 g/cm³ (20 °C) |
MDL Number:
MFCD00003413 CAS číslo: 7783-99-5 EINECS: 232-041-7 UN: 2627 ADR: 5.1,II Merck Index: 12,08663 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...