Tisk…

Silver nitrite, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Argentous nitrite

M-Clarity™ quality level = MQ100

227188-10G 227188-50G
SIAL227188-10GEA 2310 CZK
SIAL227188-10G SIAL227188-50G
Silver nitrite, Sigma-Aldrich®
Silver nitrite
Vzorec: AgNO₂
MW: 153,87 g/mol
Bod tání: 140 °C
Hustota: 4,45 g/cm³ (20 °C)
MDL Number: MFCD00003413
CAS číslo: 7783-99-5
EINECS: 232-041-7
UN: 2627
ADR: 5.1,II
Merck Index: 12,08663

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR