Tisk…

Jodid stříbrný ≥99.999% (metals basis), Premion®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Argentous iodide, Silver monoiodide

12111.06 12111.14 12111.22
12111.06EA 1620 CZK
12111.06 12111.14 12111.22
Jodid stříbrný ≥99.999% (metals basis), Premion®
Jodid stříbrný

Purity: 99.999% CAS No.: 7783-96-2 Molecular Formula: AgI Formula Weight: 234.77 Physical Form: Powder Density: 5.67 Appearance: Yellow Odour: Odourless Melting Point: 558° Merck Reference: 14,8516 UN No.: 3077 MDL No.: MFCD00003412

Vzorec: AgI
MW: 234.77
Bod varu: 1506
Bod tání: 558 °C
Hustota: 5.68
Storage Temperature: Ambient
MDL Number: MFCD00003412
CAS číslo: 7783-96-2
EINECS: 232-038-0
UN: 3077
ADR: 9 III
Merck Index: 14,8516

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR