Tisk…

Jodid stříbrný ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Argentous iodide, Silver monoiodide

11419.14 11419.22
11419.14EA 7210 CZK
11419.14 11419.22
Jodid stříbrný ≥99.9% (metals basis)
Jodid stříbrný

Purity: 99.9% CAS No.: 7783-96-2 Molecular Formula: AgI Formula Weight: 234.77 Physical Form: Crystalline Density: 5.67 Appearance: Yellow Odour: Odourless Melting Point: 558° Solubility: Insoluble in water, acids, ammonium carbonate. Freely soluble in alkali cyanides and iodides Merck Reference: 14,8516 UN No.: 3077 MDL No.: MFCD00003412

Vzorec: AgI
Bod varu: 1506 °C (1013 hPa)
Bod tání: 558 °C
Hustota: 5,67
Storage Temperature: Ambient
MDL Number: MFCD00003412
CAS číslo: 7783-96-2
EINECS: 232-038-0
UN: 3077
ADR: 9,III
Merck Index: 14,8516

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR