Tisk…

Copper(II) fluoride, anhydrous ≥99.5%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Cupric fluoride

11489.09 11489.18
11489.09EA 2620 CZK
11489.09 11489.18
Copper(II) fluoride, anhydrous ≥99.5%
Copper(II) fluoride

Purity: 99.5% CAS No.: 7789-19-7 Molecular Formula: CuF2 Formula Weight: 101.54 Physical Form: Powder Density: 4.85 Appearance: White Odour: Odourless Melting Point: 950° dec. Solubility: Solubility in water at 20° ca 4.7g/100ml Merck Reference: 14,2638 UN No.: 3260 MDL No.: MFCD00010977


Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent

Vzorec: CuF₂
MW: 101.54
Bod varu: 950
Bod tání: 950 °C (rozkl.)
Hustota: 4.85
Storage Temperature: Ambient
MDL Number: MFCD00010977
CAS číslo: 7789-19-7
EINECS: 232-147-3
UN: 3260
ADR: 8 III
Merck Index: 14,2638

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR