Copper(II) fluoride, anhydrous 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Cupric fluoride
203370250
203371000
ACRO203370250EA
5410
CZK
ACRO203370250
ACRO203371000
Copper(II) fluoride, anhydrous 98%
Copper(II) fluoride
Vzorec:
CuF₂ Bod varu: 950 °C (1013 hPa) Hustota: 4,23 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010977 CAS číslo: 7789-19-7 EINECS: 232-147-3 UN: 3260 ADR: 8,II Merck Index: 12,02704 |
Výsledky specifikačního testu
Appearance | Light grey to blue-grey Powder |
Assay | ≥97.5 % |
Loss on drying | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...