Tisk…

Uhličitan stříbrný ≥99.5% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

11420.14 11420.22
11420.14EA 5320 CZK
11420.14 11420.22
Uhličitan stříbrný ≥99.5% (metals basis)
Uhličitan stříbrný

Purity: 99.5% CAS No.: 534-16-7 Molecular Formula: CAg2O3 Formula Weight: 275.75 Physical Form: Powder Density: 6.08 Appearance: Yellow Odour: Odourless Melting Point: 218° dec. Solubility: Insoluble in water. Freely soluble in dilute HNO3, ammonia Merck Reference: 14,8507 UN No.: 3077 MDL No.: MFCD00003403

Vzorec: CAg₂O₃
MW: 275.75
Bod tání: 218 °C (rozkl.)
Hustota: 6.08
Storage Temperature: Ambient
MDL Number: MFCD00003403
CAS číslo: 534-16-7
EINECS: 208-590-3
UN: 3077
ADR: 9 III
Merck Index: 14,8507

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR