Silver perchlorate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
674583-25G
674583-5G
SIAL674583-25GEA
7760
CZK
SIAL674583-25G
SIAL674583-5G
Silver perchlorate, Sigma-Aldrich®
Silver perchlorate
Vzorec:
AgClO₄ Bod tání: 486 °C Hustota: 1,688 g/cm³ (25 °C) |
MDL Number:
MFCD00003400 CAS číslo: 7783-93-9 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II Merck Index: 12,08667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...