Tisk…

1,4-Dimethylnaphthalene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

D170305-25G D170305-5G
SIALD170305-25GEA 14330 CZK
SIALD170305-25G SIALD170305-5G
1,4-Dimethylnaphthalene, Sigma-Aldrich®
1,4-Dimethylnaphthalene
Vzorec: C₁₂H₁₂
MW: 156,23 g/mol
Bod varu: 264 °C (1013 hPa)
Hustota: 1,016 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004037
CAS číslo: 571-58-4
EINECS: 209-335-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR