2-methylnaftalen, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
β-Methylnaphthalene
M-Clarity™ quality level = MQ100
M57006-100G
M57006-500G
SIALM57006-100GEA
770
CZK
SIALM57006-100G
SIALM57006-500G
2-methylnaftalen, Sigma-Aldrich®
2-methylnaftalen
Vzorec:
C₁₁H₁₀ MW: 142,2 g/mol Bod varu: 241 °C (1013 hPa) Bod tání: 32…35 °C Hustota: 1,0058 g/cm³ (20 °C) Bod vzplanutí: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004118 CAS číslo: 91-57-6 EINECS: 202-078-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...