2-(Bromomethyl)naphthalene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
143677-100G
143677-25G
143677-5G
SIAL143677-100GEA
14860
CZK
SIAL143677-100G
SIAL143677-25G
SIAL143677-5G
2-(Bromomethyl)naphthalene, Sigma-Aldrich®
2-(Bromomethyl)naphthalene
Vzorec:
C₁₁H₉Br Bod varu: 293 °C (1013 hPa) Bod tání: 60…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004123 CAS číslo: 939-26-4 EINECS: 213-359-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...