Tisk…

2,7-Dihydroxynaphthalene ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol

A10229.18 A10229.30 A10229.36
A10229.18EA 1760 CZK
A10229.18 A10229.30 A10229.36
2,7-Dihydroxynaphthalene ≥97%
2,7-Dihydroxynaphthalene

Purity: 97% CAS No.: 582-17-2 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 183-188° Beilstein Reference: 2042383

Vzorec: C₁₀H₈O₂
Bod varu: 375 °C (1013 hPa)
Bod tání: 183…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS číslo: 582-17-2
EINECS: 209-478-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR