1,6-Dihydroxynaphthalene ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Naphthalene-1,6-diol
L15801.14
L15801.22
L15801.36
L15801.14EA
1600
CZK
L15801.14
L15801.22
L15801.36
1,6-Dihydroxynaphthalene ≥97%
1,6-Dihydroxynaphthalene
Purity: 97+%
CAS No.: 575-44-0
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 136-140°
Beilstein Reference: 1939032
Vzorec:
C₁₀H₈O₂ Bod varu: 375 °C (1013 hPa) Bod tání: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003981 CAS číslo: 575-44-0 EINECS: 209-386-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...