1-Methoxynaphthalene ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Methyl 1-naphthyl ether
B24825.14
B24825.22
B24825.36
B24825.14EA
1420
CZK
B24825.14
B24825.22
B24825.36
1-Methoxynaphthalene ≥98%
1-Methoxynaphthalene
Purity: 98+%
CAS No.: 2216-69-5
Molecular Formula: C11H10O
Formula Weight: 158.20
Flash Point:
Vzorec:
C₁₁H₁₀O MW: 158,2 g/mol Bod varu: 135…137 °C (12 mmHg) Bod tání: 5 °C Hustota: 1,094 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003924 CAS číslo: 2216-69-5 EINECS: 218-696-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...