2-Naphthoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
180246-25G
180246-5G
SIAL180246-25GEA
2050
CZK
SIAL180246-25G
SIAL180246-5G
2-Naphthoic acid, Sigma-Aldrich®
2-Naphthoic acid
Vzorec:
C₁₁H₈O₂ Bod varu: 333 °C (1013 hPa) Bod tání: 180…183 °C Hustota: 1,382 g/cm³ (20 °C) |
MDL Number:
MFCD00004101 CAS číslo: 93-09-4 EINECS: 202-217-8 Merck Index: 12,06469 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...