Tisk…

1-Naphthyl boronic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

M-Clarity™ quality level = MQ100

N257-25G N257-5G
SIALN257-25GEA 8680 CZK
SIALN257-25G SIALN257-5G
1-Naphthyl boronic acid, Sigma-Aldrich®
1-Naphthyl boronic acid
Vzorec: C₁₀H₉BO₂
MW: 171,99 g/mol
Bod tání: 259 °C
MDL Number: MFCD00019722
CAS číslo: 13922-41-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR