Tisk…

1-Naphthyl boronic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

B21219.03 B21219.06 B21219.14
B21219.03EA 1430 CZK
B21219.03 B21219.06 B21219.14
1-Naphthyl boronic acid ≥97%
1-Naphthyl boronic acid

Purity: 97% CAS No.: 13922-41-3 Molecular Formula: C10H9BO2 Formula Weight: 171.99 Melting Point: 204-210° Beilstein Reference: 2937504

Vzorec: C₁₀H₉BO₂
MW: 171,99 g/mol
Bod tání: 204…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00019722
CAS číslo: 13922-41-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR