Tisk…

Lanthanum(III) sulphate, anhydrous ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Dilanthanum trisulphate

89760.18 89760.30
89760.18EA 3880 CZK
89760.18 89760.30
Lanthanum(III) sulphate, anhydrous ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) sulphate

Purity: 99.9% CAS No.: 10099-60-2 Molecular Formula: La2(SO4)3 Formula Weight: 566.00 Physical Form: Powder Density: 3.60 Appearance: White Odour: Odourless Melting Point: 1150° dec. Merck Reference: 14,5363 MDL No.: MFCD00011070

Vzorec: La₂O₁₂S₃
MW: 566
Bod tání: 1150 °C (rozkl.)
Hustota: 3.60
Storage Temperature: Ambient
MDL Number: MFCD00011070
CAS číslo: 10099-60-2
EINECS: 233-239-6
Merck Index: 14,5363

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR