Cerium(IV) sulphate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
359009-100G
359009-25G
SIAL359009-100GEA
4380
CZK
SIAL359009-100G
SIAL359009-25G
Cerium(IV) sulphate, Sigma-Aldrich®
Cerium(IV) sulphate
Vzorec:
Ce(SO₄)₂ Bod tání: 195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00148852 CAS číslo: 13590-82-4 EINECS: 237-029-5 UN: 3260 ADR: 8,III Merck Index: 12,02036 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...