Tisk…

Lanthanum(III) hydroxide ≥99.95%

Dodavatel: Thermo Fisher Scientific
Varování

43123.18 43123.30
43123.18EA 2430 CZK
43123.18 43123.30
Lanthanum(III) hydroxide ≥99.95%
Lanthanum(III) hydroxide

Purity: 99.95% CAS No.: 14507-19-8 Molecular Formula: La(OH)3 Formula Weight: 189.93 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: dec. MDL No.: MFCD00016160

Vzorec: H₃LaO₃
MW: 189.93
Bod tání: Dec.
Hustota: 4.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016160
CAS číslo: 14507-19-8
EINECS: 238-510-2
UN: 3262
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR