Oxid ytterbitý ≥99.9% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Fisher Scientific
Synonyms:
Diytterbium trioxide, Ytterbium oxide
11191.14
11191.22
11191.A1
11191.14EA
3230
CZK
11191.14
11191.22
11191.A1
Oxid ytterbitý ≥99.9% (REO, rare earth oxide basis), REacton®
Oxid ytterbitý
Purity: 99.9%
CAS No.: 1314-37-0
Molecular Formula: Yb2O3
Formula Weight: 394.08
Physical Form: Powder
Density: 9.2
Melting Point: 2227°
Merck Reference: 14,10105
MDL No.: MFCD00011290
Vzorec:
O₃Yb₂ MW: 394.08 Bod tání: 2227 °C Hustota: 9.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011290 CAS číslo: 1314-37-0 EINECS: 215-234-0 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...