Tisk…
Safety Shoes, sandals, Velcro® fastening, Basic, Leon, Jori
Dodavatel: ELTEN
11401/36 11401/37 11401/38 11401/39 11401/40 11401/41 11401/42 11401/43 11401/44 11401/45 11401/46 11401/47 11401/48
ELTN11401/36EA 1290 CZK
ELTN11401/36 ELTN11401/37 ELTN11401/38 ELTN11401/39 ELTN11401/40 ELTN11401/41 ELTN11401/42 ELTN11401/43 ELTN11401/44 ELTN11401/45 ELTN11401/46 ELTN11401/47 ELTN11401/48
Safety Shoes, sandals, Velcro® fastening, Basic, Leon, Jori
Obuv
Durable safety sandals made from wear resistant cowhide leather, skin-friendly textile lining, a full padded bellows tongue and collar padding and a dual-density PU/PU NEW BASIC sole (oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in dryMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí ISO 14001