Tisk…

Oxid tulitý ≥99.99% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

11199.04 11199.09
11199.04EA 6760 CZK
11199.04 11199.09
Oxid tulitý ≥99.99% (REO, rare earth oxide basis), REacton®
Oxid tulitý

Purity: 99.99% CAS No.: 12036-44-1 Molecular Formula: O3Tm2 Formula Weight: 385.87 Physical Form: Powder Appearance: Pale green Melting Point: 2425° Merck Reference: 14,9394 MDL No.: MFCD00011285

Vzorec: O₃Tm₂
MW: 385.87
Bod tání: 2425 °C
Hustota: 8.6
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS číslo: 12036-44-1
EINECS: 234-851-6
Merck Index: 14,9394

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR