Tisk…

Iron(II) oxide ≥99.7% (trace metals basis), powder ~ 10 mesh, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

400866-25G 400866-5G
SIAL400866-25GEA 9960 CZK
SIAL400866-25G SIAL400866-5G
Iron(II) oxide ≥99.7% (trace metals basis), powder ~ 10 mesh, Sigma-Aldrich®
Iron(II) oxide

Employed in the synthesis of the first example of a mixed-valence ammonium iron phosphate, NH₄Fe₂(PO₄)₂.

Vzorec: FeO
Hustota: 5,7 g/ml (25 °C)
MDL Number: MFCD00016095
CAS číslo: 1345-25-1
EINECS: 215-721-8

Objednat

Výsledky specifikačního testu

Purity (Trace metals basis) 99.7%
Fe (Titration by Na₂S₂O₃) 76.2 - 79.3%
Iron component (ICP major analysis) Confirmed
Trace metal analysis <3500.0 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR