Oxid samaritý ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
36278.09
36278.18
36278.09EA
3710
CZK
36278.09
36278.18
Oxid samaritý ≥99.9% (metals basis)
Oxid samaritý
Purity: 99.9%
CAS No.: 12060-58-1
Molecular Formula: Sm2O3
Formula Weight: 348.70
Physical Form: 3-12mm Sintered Lump
Refractive Index: 1.97
Melting Point: 2300°
Merck Reference: 14,8347
MDL No.: MFCD00011237
Vzorec:
O₃Sm₂ MW: 348.72 Bod tání: 2300 °C Hustota: 7.68 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011237 CAS číslo: 12060-58-1 EINECS: 235-043-6 Merck Index: 14,8347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...