Tisk…

Oxid samaritý ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Scientific

11229.14 11229.22 11229.36 11229.A1
11229.14EA 1680 CZK
11229.14 11229.22 11229.36 11229.A1
Oxid samaritý ≥99.9% (REO, rare earth oxide basis), REacton®
Oxid samaritý

Purity: 99.9% CAS No.: 12060-58-1 Molecular Formula: Sm2O3 Formula Weight: 348.70 Physical Form: Powder Refractive Index: 1.97 Melting Point: 2300° Merck Reference: 14,8347 MDL No.: MFCD00011237

Vzorec: O₃Sm₂
MW: 348,72 g/mol
Bod tání: 2300 °C
Hustota: 7,68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011237
CAS číslo: 12060-58-1
EINECS: 235-043-6
Merck Index: 14,8347

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR