Oxid neodymitý, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
M-Clarity™ quality level = MQ100
228656-100G
228656-25G
SIAL228656-100GEA
10080
CZK
SIAL228656-100G
SIAL228656-25G
Oxid neodymitý, Sigma-Aldrich®
Oxid neodymitý
Vzorec:
Nd₂O₃ MW: 336.48 Bod varu: 3760 Bod tání: 2270 Hustota: 7.24 |
MDL Number:
MFCD00011134 CAS číslo: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,06450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...