Tisk…

Oxid neodymitý, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

M-Clarity™ quality level = MQ100

203858-10G 203858-50G
SIAL203858-10GEA 5750 CZK
SIAL203858-10G SIAL203858-50G
Oxid neodymitý, Sigma-Aldrich®
Oxid neodymitý
Vzorec: Nd₂O₃
MW: 336.48
Bod varu: 3760
Bod tání: 2270
Hustota: 7.24
MDL Number: MFCD00011134
CAS číslo: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR