Tisk…

Oxid gadolinitý ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Gadolinium (III) oxide

11291.14 11291.22 11291.36
11291.14EA 2670 CZK
11291.14 11291.22 11291.36
Oxid gadolinitý ≥99.9% (REO, rare earth oxide basis), REacton®
Oxid gadolinitý

Purity: 99.9% CAS No.: 12064-62-9 Molecular Formula: Gd2O3 Formula Weight: 362.50 Physical Form: 10 Micron Powder Density: 7.407 Melting Point: 2330° Merck Reference: 14,4326 MDL No.: MFCD00011026

Vzorec: Gd₂O₃
MW: 362.5
Bod tání: 2330 °C
Hustota: 7.407
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS číslo: 12064-62-9
EINECS: 235-060-9
Merck Index: 14,4326

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR