Tisk…

Oxid ceričitý ≥99.5% (REO, rare earth oxide basis), nanopowder

Dodavatel: Thermo Fisher Scientific
Varování

44960.36 44960.22 44960.14
44960.36EA 27110 CZK
44960.36 44960.22 44960.14
Oxid ceričitý ≥99.5% (REO, rare earth oxide basis), nanopowder
Oxid ceričitý

Purity: 99.5% min CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: 15-30nm APS Powder, S.A. 30-50 m2/g Density: 7.132 Appearance: Pale yellow Melting Point: 2600° Merck Reference: 14,1989 MDL No.: MFCD00010933

Vzorec: CeO₂
Bod varu: 3468 °C (1013 hPa)
Bod tání: 2600 °C
Hustota: 7,132
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS číslo: 1306-38-3
EINECS: 215-150-4
Merck Index: 14,1989

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR