Tisk…

Thulium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Varování


Ampuled under argon

39424.03 39424.06
39424.03EA 3290 CZK
39424.03 39424.06
Thulium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Thulium(III) iodide

Purity: 99.99% CAS No.: 13813-43-9 Molecular Formula: I3Tm Formula Weight: 549.65 Physical Form: Flakes, Ampouled under argon Sensitivity: Hygroscopic Boiling Point: 1260° MDL No.: MFCD00050141

Vzorec: I₃Tm
Bod varu: 1260 °C
Bod tání: 1015 °C
Storage Temperature: Ambient
MDL Number: MFCD00050141
CAS číslo: 13813-43-9
EINECS: 237-472-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR