Tisk…

Lanthanum(III) iodide, ultra dry ≥99.995% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Lanthanum triiodide

Ampuled under argon

44548.03 44548.06
44548.03EA 4640 CZK
44548.03 44548.06
Lanthanum(III) iodide, ultra dry ≥99.995% (metals basis)
Lanthanum(III) iodide

Purity: 99.995+% CAS No.: 13813-22-4 Molecular Formula: LaI3 Formula Weight: 519.62 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Moisture Sensitive MDL No.: MFCD00049470

Vzorec: I₃La
MW: 519.62
Bod tání: 772 °C
Hustota: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049470
CAS číslo: 13813-22-4
EINECS: 237-465-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR