Holmium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Ampuled under argon
13635.03
13635.06
13635.03EA
4640
CZK
13635.03
13635.06
Holmium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Holmium triiodide
Purity: 99.99%
CAS No.: 13813-41-7
Molecular Formula: HoI3
Formula Weight: 545.64
Physical Form: Flakes, Ampouled under argon
Sensitivity: Hygroscopic
Merck Reference: 14,4727
MDL No.: MFCD00049745
Vzorec:
HoI₃ MW: 545,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049745 CAS číslo: 13813-41-7 EINECS: 237-470-3 Merck Index: 14,4727 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...