Tisk…

Gadolinium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Varování


Ampuled under argon

35735.03 35735.06
35735.03EA 4740 CZK
35735.03 35735.06
Gadolinium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Gadolinium triiodide

Purity: 99.99% CAS No.: 13572-98-0 Molecular Formula: GdI3 Formula Weight: 537.96 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00016114

Vzorec: GdI₃
Bod tání: 926 °C
Storage Temperature: Ambient
MDL Number: MFCD00016114
CAS číslo: 13572-98-0
EINECS: 236-997-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR