Dysprosium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Ampuled under argon
35693.03
35693.06
35693.03EA
6730
CZK
35693.03
35693.06
Dysprosium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Dysprosium triiodide
Purity: 99.99%
CAS No.: 15474-63-2
Molecular Formula: DyI3
Formula Weight: 543.21
Physical Form: Flake, Ampouled under argon
Sensitivity: Hygroscopic
Appearance: Green to yellow
Melting Point: 955°
MDL No.: MFCD00016072
Vzorec:
DyI₃ MW: 543.21 Bod tání: 955 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016072 CAS číslo: 15474-63-2 EINECS: 239-493-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...