Thulium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
13652.06
13652.14
13652.06EA
8790
CZK
13652.06
13652.14
Thulium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Thulium(III) fluoride
Purity: 99.9%
CAS No.: 13760-79-7
Molecular Formula: TmF3
Formula Weight: 225.93
Physical Form: Crystalline
Density: 7.9
Melting Point: 1158°
Boiling Point: 2200°
Solubility: Insoluble in water
UN No.: 3288
MDL No.: MFCD00049611
Vzorec:
F₃Tm MW: 225,93 g/mol Bod varu: 2200 °C Bod tání: 1158 °C Hustota: 7,9 Storage Temperature: Ambient |
MDL Number:
MFCD00049611 CAS číslo: 13760-79-7 EINECS: 237-353-7 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...