Samarium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
11226.04
11226.09
11226.18
11226.04EA
1660
CZK
11226.04
11226.09
11226.18
Samarium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium(III) fluoride
Purity: 99.99%
CAS No.: 13765-24-7
Molecular Formula: SmF3
Formula Weight: 207.35
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 3288
MDL No.: MFCD00049540
Vzorec:
F₃Sm Bod varu: 2323 °C (1013 hPa) Bod tání: 1306 °C Hustota: 4,362 g/cm³ (2327 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049540 CAS číslo: 13765-24-7 EINECS: 237-367-3 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...