Tisk…

Neodymium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

13661.14 13661.22
13661.14EA 4400 CZK
13661.14 13661.22
Neodymium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium trifluoride

Purity: 99.9% CAS No.: 13709-42-7 Molecular Formula: NdF3 Formula Weight: 201.24 Physical Form: Crystalline Lump Density: 6.5 Appearance: Pale purple Odour: Odourless Melting Point: 1410° MDL No.: MFCD00011135

Vzorec: F₃Nd
Bod varu: 2327 °C (1013 hPa)
Bod tání: 1410 °C
Hustota: 6,5
Storage Temperature: Ambient
MDL Number: MFCD00011135
CAS číslo: 13709-42-7
EINECS: 237-253-3
UN: 3288
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR