Tisk…

Lutetium trifluoride ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Holmium(III) fluoride, Lutetium (III) fluoride

44425.03 44425.06
44425.03EA 4120 CZK
44425.03 44425.06
Lutetium trifluoride ≥99.9% (metals basis)
Lutetium trifluoride

Purity: 99.9% CAS No.: 13760-81-1 Molecular Formula: LuF3 Formula Weight: 231.97 Physical Form: -325 Mesh Powder Density: 8.3 Melting Point: 1182° UN No.: 3288 MDL No.: MFCD00016191

Vzorec: LuF₃
Bod varu: 2200 °C (1013 hPa)
Bod tání: 1182 °C
Hustota: 8,3
Storage Temperature: Ambient
MDL Number: MFCD00016191
CAS číslo: 13760-81-1
EINECS: 237-355-8
UN: 3288
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR