Tisk…

Lanthanum(III) fluoride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Lanthanum trifluoride

M-Clarity™ quality level = MQ100

449857-25G 449857-5G
SIAL449857-25GEA 17060 CZK
SIAL449857-25G SIAL449857-5G
Lanthanum(III) fluoride, Sigma-Aldrich®
Lanthanum(III) fluoride
Vzorec: F₃La
MW: 195.9
Bod varu: ∼2327 °C (1013 hPa)
Bod tání: 1491.85…1507.85 °C
Hustota: 5.936 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011073
CAS číslo: 13709-38-1
EINECS: 237-252-8
UN: 3288
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR