Lanthanum(III) fluoride ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Lanthanum trifluoride
A16673.14
A16673.22
A16673.14EA
1830
CZK
A16673.14
A16673.22
Lanthanum(III) fluoride ≥99%
Lanthanum(III) fluoride
Purity: 99%
CAS No.: 13709-38-1
Molecular Formula: F3La
Formula Weight: 195.90
Odour: Odourless
Melting Point: 1493°
MDL No.: MFCD00011073
Vzorec:
F₃La Bod varu: ∼2327 °C (1013 hPa) Bod tání: 1493 °C Hustota: 5,936 Storage Temperature: Ambient |
MDL Number:
MFCD00011073 CAS číslo: 13709-38-1 EINECS: 237-252-8 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...