Tisk…

Holmium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Holmium (III) fluoride

45033.18 45033.09
45033.18EA 17540 CZK
45033.18 45033.09
Holmium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Holmium trifluoride

Purity: 99.9% CAS No.: 13760-78-6 Molecular Formula: HoF3 Formula Weight: 221.93 Physical Form: Powder Density: 7.6 Melting Point: 1143° UN No.: 3288 MDL No.: MFCD00016142

Vzorec: HoF₃
Bod tání: 1143 °C
Hustota: 7,6
Storage Temperature: Ambient
MDL Number: MFCD00016142
CAS číslo: 13760-78-6
EINECS: 237-352-1
UN: 3288
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR