Holmium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Holmium (III) fluoride
41413.09
41413.04
41413.09EA
5080
CZK
41413.09
41413.04
Holmium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Holmium trifluoride
Purity: 99.9%
CAS No.: 13760-78-6
Molecular Formula: HoF3
Formula Weight: 221.93
Physical Form: 3-6mm Pieces
Density: 7.6
Melting Point: 1143°
UN No.: 3288
MDL No.: MFCD00016142
For vacuum deposition
Vzorec:
HoF₃ MW: 221.93 Bod tání: 1143 Hustota: 7.6 Storage Temperature: Ambient |
MDL Number:
MFCD00016142 CAS číslo: 13760-78-6 EINECS: 237-352-1 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...