Europium(II) fluoride, anhydrous ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
41737.02
41737.04
41737.09
41737.02EA
2700
CZK
41737.02
41737.04
41737.09
Europium(II) fluoride, anhydrous ≥99.9% (metals basis)
Europium(II) fluoride
Purity: 99.9%
CAS No.: 14077-39-5
Molecular Formula: EuF2
Formula Weight: 189.96
Physical Form: -200 Mesh Powder
Density: 6.495
Melting Point: 1380°
Boiling Point:
Vzorec:
EuF₂ Bod varu: >2400 °C Bod tání: 1380 °C Hustota: 6,495 Storage Temperature: Ambient |
MDL Number:
MFCD01866333 CAS číslo: 14077-39-5 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...