Tisk…

Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

13656.14 13656.22
13656.14EA 3540 CZK
13656.14 13656.22
Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium trifluoride

Purity: 99.9% CAS No.: 13569-80-7 Molecular Formula: DyF3 Formula Weight: 219.50 Physical Form: Crystalline Lump Density: 7.5 Appearance: White Odour: Odourless Melting Point: 1360° Boiling Point: 2200° MDL No.: MFCD00016071

Vzorec: DyF₃
Bod varu: 2200 °C
Bod tání: 1360 °C
Hustota: 7,5
Storage Temperature: Ambient
MDL Number: MFCD00016071
CAS číslo: 13569-80-7
EINECS: 236-992-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR