Cerium trifluoride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cerous fluoride, Cerium (III) fluoride
M-Clarity™ quality level = MQ100
229555-10G
229555-50G
SIAL229555-10GEA
7010
CZK
SIAL229555-10G
SIAL229555-50G
Cerium trifluoride, Sigma-Aldrich®
Cerium trifluoride
Vzorec:
CeF₃ MW: 197.11 Bod varu: ∼1730 °C (1013 hPa) Bod tání: 817 Hustota: 3.92 g/cm³ (20 °C) |
MDL Number:
MFCD00010932 CAS číslo: 7758-88-5 EINECS: 231-841-3 Merck Index: 12,02042 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...