Ytterbium(III) chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
450073-25G
450073-5G
SIAL450073-25GEA
22460
CZK
SIAL450073-25G
SIAL450073-5G
Ytterbium(III) chloride, Sigma-Aldrich®
Ytterbium(III) chloride
Vzorec:
Cl₃Yb MW: 279,4 g/mol Bod tání: 860 °C |
MDL Number:
MFCD00011288 CAS číslo: 10361-91-8 EINECS: 233-800-5 Merck Index: 13,10014 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...