Ytterbium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Ampuled under argon
40653.06
40653.14
40653.06EA
6760
CZK
40653.06
40653.14
Ytterbium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Ytterbium(III) chloride
Purity: 99.99%
CAS No.: 10361-91-8
Molecular Formula: YbCl3
Formula Weight: 279.40
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Hygroscopic
Merck Reference: 14,10105
MDL No.: MFCD00011288
Vzorec:
Cl₃Yb MW: 279.4 Bod tání: 854 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011288 CAS číslo: 10361-91-8 EINECS: 233-800-5 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...