Terbium(III) chloride hexahydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
204560-1G
204560-5G
SIAL204560-1GEA
3430
CZK
SIAL204560-1G
SIAL204560-5G
Terbium(III) chloride hexahydrate, Sigma-Aldrich®
Terbium(III) chloride hexahydrate
Vzorec:
Cl₃Tb·6H₂O MW: 373.38 Bod tání: 145 Hustota: 4.35 g/cm³ (20 °C) |
MDL Number:
MFCD00149870 CAS číslo: 13798-24-8 EINECS: 233-132-4 Merck Index: 12,09300 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...