Lutetium trichloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
450960-1G
450960-5G
SIAL450960-1GEA
6950
CZK
SIAL450960-1G
SIAL450960-5G
Lutetium trichloride, Sigma-Aldrich®
Lutetium trichloride
Vzorec:
Cl₃Lu MW: 281,33 g/mol Bod tání: 892 °C Hustota: 3,98 g/cm³ (25 °C) |
MDL Number:
MFCD00011099 CAS číslo: 10099-66-8 EINECS: 233-240-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...